siRNA Details
| Gene Symbol: Gm15894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038870.1 | - | 262 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632619.1 | - | 2 | 259 | Chr. 8 - 110754994 - 110757586 [+] on Build GRCm38 | |

Gm15894
Genomic Map