siRNA Details
| Gene Symbol: Bcas3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045875.1 | 2 | 2394 | |||
| GenBank mRNA | AK018376.1 | - | 1, 2 | 60 | ||
| AK053124.1 | - | 2 | 2395 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720493.1 | - | 1, 2 | 60 | Chr. 11 - 85704289 - 85719744 [-] on Build GRCm38 | |

Bcas3os1
Genomic Map