siRNA Details
Gene Symbol: Bcas3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045875.1 | 4 | 2637 | |||
GenBank mRNA | AK018376.1 | - | 4 | 468 | ||
AK053124.1 | - | 4 | 2638 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bcas3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045875.1 | 4 | 2637 | |||
GenBank mRNA | AK018376.1 | - | 4 | 468 | ||
AK053124.1 | - | 4 | 2638 |