siRNA Details
| Gene Symbol: Bcas3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045875.1 | 3 | 2602 | |||
| GenBank mRNA | AK018376.1 | - | 3 | 268 | ||
| AK053124.1 | - | 3 | 2603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcas3os1| Gene Symbol: Bcas3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045875.1 | 3 | 2602 | |||
| GenBank mRNA | AK018376.1 | - | 3 | 268 | ||
| AK053124.1 | - | 3 | 2603 | |||