siRNA Details
| Gene Symbol: Bcas3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045875.1 | 5 | 2738 | |||
| GenBank mRNA | AK018376.1 | - | 5 | 569 | ||
| AK053124.1 | - | 5 | 2739 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB530090.2 | - | 4 | 293 | Chr. 11 - 85702200 - 85706817 [-] on Build GRCm38 | |

Bcas3os1
Genomic Map