siRNA Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138554.1 | - | 2 | 187 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB633564.1 | - | 2 | 187 | Chr. 1 - 21166498 - 21187631 [+] on Build GRCm38 | |

Tmem14a
Genomic Map