siRNA Details
| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020409.1 | - | 1 | 264 | ||
| AK145503.1 | - | 1 | 2079 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx20| Gene Symbol: Tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020409.1 | - | 1 | 264 | ||
| AK145503.1 | - | 1 | 2079 | |||