siRNA Details
| Gene Symbol: Gm38477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077098.1 | - | 2 | 609 | ||
| AK133303.1 | - | 2415 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616944.1 | - | 2 | 609 | Chr. 16 - 87004267 - 87023797 [-] on Build GRCm38 | |

Gm38477
Genomic Map