siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017359.1 | - | 1 | 614 | ||
| AK049081.1 | - | 1 | 681 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ammecr1l| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017359.1 | - | 1 | 614 | ||
| AK049081.1 | - | 1 | 681 | |||