siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017359.1 | - | 1 | 203 | ||
| AK049081.1 | - | 1 | 266 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG861658.1 | - | 1 | 31 | Chr. 18 - 31782749 - 31783538 [+] on Build GRCm38 | |

Ammecr1l
Genomic Map