siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017601.1 | - | 1 | 243 | ||
| AK021120.1 | - | 1 | 422 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728248.1 | - | 1 | 420 | Chr. 18 - 69592604 - 69593075 [+] on Build GRCm38 | |

Tcf4
Genomic Map