siRNA Details
| Gene Symbol: Hoxd3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135513.1 | - | 1 | 387 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052125.1 | - | 1 | 387 | Chr. 2 - 74713050 - 74713697 [+] on Build GRCm38 | |

Hoxd3os1
Genomic Map