siRNA Details
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052877.1 | - | 1 | 672 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB516668.2 | - | 1 | 31 | Chr. 19 - 30564489 - 30565147 [-] on Build GRCm38 |
