siRNA Details
| Gene Symbol: 9530020I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020565.1 | - | 3 | 633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530020I12Rik| Gene Symbol: 9530020I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020565.1 | - | 3 | 633 | ||