siRNA Details
| Gene Symbol: Psg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030357.1 | - | 6 | 3297 | 5367680 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK077897.1 | - | 90 | Chr. - - [-] on Build | ||

Psg16
Genomic Map