siRNA Details
| Gene Symbol: Thap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006173.1 | - | 3 | 514 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706272.1 | - | 3 | 514 | Chr. 7 - 30280094 - 30289704 [+] on Build GRCm38 | |

Thap8
Genomic Map