siRNA Details
| Gene Symbol: C330008A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021191.1 | - | 1 | 56 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728756.1 | - | 1 | 51 | Chr. 18 - 35913135 - 35916753 [+] on Build GRCm38 | |

C330008A17Rik
Genomic Map