siRNA Details
| Gene Symbol: 1700097M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007098.1 | - | 1 | 984 | ||
| AV270744.2 | - | 1 | 93 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK076915.1 | - | 275 | Chr. - - [-] on Build | ||

1700097M23Rik
Genomic Map