siRNA Details
| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037997.1 | 4 | 2841 | |||
| GenBank mRNA | AK009355.1 | - | 1 | 1528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310015D24Rik| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037997.1 | 4 | 2841 | |||
| GenBank mRNA | AK009355.1 | - | 1 | 1528 | ||