siRNA Details
| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037997.1 | 4 | 1759 | |||
| GenBank mRNA | AK009355.1 | - | 1 | 445 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709122.1 | - | 1 | 443 | Chr. 16 - 13514724 - 13515728 [-] on Build GRCm38 | |

2310015D24Rik
Genomic Map