siRNA Details
Gene Symbol: Gm12100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049252.1 | - | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB651855.1 | - | 1 | 167 | Chr. 11 - 30750832 - 30754861 [+] on Build GRCm38 |
