siRNA Details
| Gene Symbol: Gm13799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029614.1 | - | 1, 2 | 125 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615000.1 | - | 1, 2 | 125 | Chr. 2 - 95838088 - 95840554 [+] on Build GRCm38 | |

Gm13799
Genomic Map