siRNA Details
| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046307.1 | 4 | 781 | |||
| GenBank mRNA | AK015475.1 | - | 4 | 783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930405A10Rik| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046307.1 | 4 | 781 | |||
| GenBank mRNA | AK015475.1 | - | 4 | 783 | ||