siRNA Details
| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015475.1 | - | 4 | 666 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715024.1 | - | 4 | 666 | Chr. 14 - 22507276 - 22542798 [-] on Build GRCm38 | |

4930405A10Rik
Genomic Map