siRNA Details
| Gene Symbol: Gm15880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040343.1 | 1, 2 | 86 | |||
| GenBank mRNA | AK155169.1 | - | 1, 2 | 87 | ||
| BY749974.1 | - | 1, 2 | 87 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY748779.1 | - | 1, 2 | 87 | Chr. 7 - 80636017 - 80642187 [+] on Build GRCm38 | |

Gm15880
Genomic Map