siRNA Details
| Gene Symbol: Gm36633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076715.1 | - | 5 | 525 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm36633| Gene Symbol: Gm36633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076715.1 | - | 5 | 525 | ||