siRNA Details
| Gene Symbol: 3110023J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014075.1 | - | 1 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110023J12Rik| Gene Symbol: 3110023J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014075.1 | - | 1 | 332 | ||