siRNA Details
Gene Symbol: G630018N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090209.1 | - | 4, 5 | 562 | ||
AK143085.1 | - | 3, 4 | 289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: G630018N14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090209.1 | - | 4, 5 | 562 | ||
AK143085.1 | - | 3, 4 | 289 |