siRNA Details
| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040684.1 | 7 | 963 | |||
| GenBank mRNA | AK016926.1 | - | 7 | 962 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkag2os1| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040684.1 | 7 | 963 | |||
| GenBank mRNA | AK016926.1 | - | 7 | 962 | ||