siRNA Details
| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040684.1 | 5, 6 | 768 | |||
| GenBank mRNA | AK016926.1 | - | 5, 6 | 767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkag2os1| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040684.1 | 5, 6 | 768 | |||
| GenBank mRNA | AK016926.1 | - | 5, 6 | 767 | ||