siRNA Details
| Gene Symbol: Gm17767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043869.1 | - | 533 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB641720.1 | - | 3 | 533 | Chr. 1 - 51582626 - 51596718 [-] on Build GRCm38 | |

Gm17767
Genomic Map