siRNA Details
Gene Symbol: Gm17767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043869.1 | - | 124 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB641720.1 | - | 1 | 124 | Chr. 1 - 51582626 - 51596718 [-] on Build GRCm38 |
