siRNA Details
| Gene Symbol: Gm19510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045076.1 | 4 | 1635 | |||
| GenBank mRNA | AK035211.1 | - | 4 | 1636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19510| Gene Symbol: Gm19510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045076.1 | 4 | 1635 | |||
| GenBank mRNA | AK035211.1 | - | 4 | 1636 | ||