siRNA Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035217.1 | - | 1 | 871 | ||
| AK080172.1 | - | 1 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam129a| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035217.1 | - | 1 | 871 | ||
| AK080172.1 | - | 1 | 367 | |||