siRNA Details
| Gene Symbol: Gm36503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144227.1 | - | 2 | 147 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY733334.1 | - | 2 | 146 | Chr. 6 - 16550387 - 16579681 [+] on Build GRCm38 | |

Gm36503
Genomic Map