siRNA Details
Gene Symbol: 2810047F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012915.1 | - | 4 | 2009 | ||
AK081830.1 | - | 1 | 1032 | |||
BB375486.2 | - | 1 | 292 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2810047F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012915.1 | - | 4 | 2009 | ||
AK081830.1 | - | 1 | 1032 | |||
BB375486.2 | - | 1 | 292 |