siRNA Details
| Gene Symbol: D630004K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052609.1 | - | 1 | 2457 | ||
| BB335455.2 | - | 1 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630004K10Rik| Gene Symbol: D630004K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052609.1 | - | 1 | 2457 | ||
| BB335455.2 | - | 1 | 588 | |||