siRNA Details
Gene Symbol: D630004K10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052609.1 | - | 1 | 97 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ228391.1 | - | 1 | 691 | Chr. 10 - 125221650 - 125222361 [-] on Build GRCm38 |
