siRNA Details
| Gene Symbol: D630004K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052609.1 | - | 1 | 1579 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF468885.1 | - | 1 | 195 | Chr. 10 - 125219969 - 125220399 [-] on Build GRCm38 | |

D630004K10Rik
Genomic Map