siRNA Details
Gene Symbol: 4933433F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045856.1 | 1 | 120 | |||
GenBank mRNA | AK017038.1 | - | 1 | 123 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716722.1 | - | 1 | 123 | Chr. 8 - 30568393 - 30574639 [+] on Build GRCm38 |
