siRNA Details
Gene Symbol: 4933433F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045856.1 | 2 | 557 | |||
GenBank mRNA | AK017038.1 | - | 2 | 560 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716722.1 | - | 2 | 560 | Chr. 8 - 30568393 - 30574639 [+] on Build GRCm38 |
