siRNA Details
| Gene Symbol: 4930588D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019831.1 | - | 1 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716010.1 | - | 1 | 54 | Chr. 15 - 80227939 - 80228414 [-] on Build GRCm38 | |

4930588D02Rik
Genomic Map