siRNA Details
| Gene Symbol: 2410021H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045428.1 | 3 | 492 | |||
| GenBank mRNA | AK010563.1 | - | 3 | 497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410021H03Rik| Gene Symbol: 2410021H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045428.1 | 3 | 492 | |||
| GenBank mRNA | AK010563.1 | - | 3 | 497 | ||