siRNA Details
| Gene Symbol: 2410021H03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045428.1 | 3 | 249 | |||
| GenBank mRNA | AK010563.1 | - | 3 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709978.1 | - | 3 | 252 | Chr. 17 - 69275378 - 69277208 [-] on Build GRCm38 | |

2410021H03Rik
Genomic Map