siRNA Details
| Gene Symbol: 4930589O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019834.1 | - | 220 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV049786.1 | - | 1 | 259 | Chr. 5 - 65493076 - 65493973 [+] on Build GRCm38 | |

4930589O11Rik
Genomic Map