siRNA Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009638.1 | - | 494 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ571170.1 | - | 1 | 515 | Chr. 5 - 108503057 - 108503797 [+] on Build GRCm38 | |

Pcgf3
Genomic Map