siRNA Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009638.1 | - | 1070 | |||
| BC049266.1 | - | 1541 | 5371040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcgf3| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009638.1 | - | 1070 | |||
| BC049266.1 | - | 1541 | 5371040 | |||