siRNA Details
| Gene Symbol: 1700039O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007166.1 | - | 1 | 203 | ||
| AK018843.1 | - | 1 | 197 | |||
| AK018940.1 | - | 1 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700039O17Rik| Gene Symbol: 1700039O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007166.1 | - | 1 | 203 | ||
| AK018843.1 | - | 1 | 197 | |||
| AK018940.1 | - | 1 | 198 | |||