siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007167.1 | - | 1 | 265 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707251.1 | - | 1 | 265 | Chr. 7 - 80246425 - 80246806 [+] on Build GRCm38 | |

Ttll13
Genomic Map