siRNA Details
| Gene Symbol: 1700110K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040728.1 | 3 | 656 | |||
| GenBank mRNA | AK007162.1 | - | 3 | 659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700110K17Rik| Gene Symbol: 1700110K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040728.1 | 3 | 656 | |||
| GenBank mRNA | AK007162.1 | - | 3 | 659 | ||